Order Entry
Switzerland
ContactUsLinkComponent
Glass fibre filters, without binder
Glass fibre filters, without binder
  PRATA0258837
undefined
Glass fibre filters, without binder
  PRATA0258837

 

  • Grade:
    GF/A
  • Size:
    25 mm
  • Pore size:
    1,2 µm
  • Pk:
    100

 

 

Borosilicate glass fibre filters without binder.

  • Pure quality in white colour
  • For use in very high-temperature (380 °C)