Technische Daten
- Antibody type:Primary
- Antigen name:Echinoderm microtubule associated Protein like 3
- Antigen symbol:EML3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:256364
- Antigen synonyms:ELP95
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Technische Daten
Über diesen Artikel
At the onset of mitosis, assembly of the mitotic spindle requires a global change in the activity of microtubule-binding proteins. EML3 (Echinoderm microtubule-associated protein-like 3) is a 896 amino acid protein that likely modifies microtubule dynamics by making them longer. Through colocalization with spindle microtubules during mitosis, EML3 plays a role in correct metaphase chromosome alignment. EML3 contains a nuclear localization signal and a microtubule-binding domain. The gene encoding EML3 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: EML3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat