- Antibody type:Primary
- Antigen name:Hairy/enhancer-of-split related with YRPW motif 2
- Antigen symbol:HEY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:23493
- Antigen synonyms:GRL|bHLHb32|GRIDLOCK|HESR2|HRT2|HERP1|CHF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
The LIN-12/Notch family of transmembrane receptors plays a central role in development by regulating cell fate and establishing boundaries of gene expression. Notch signaling activates the Hairy/Enhancer of split (HES) genes, which encode basic helix-loop-helix (bHLH) transcriptional repressors that are critical for directing embryonic patterning and development. The Hairy-related transcription factors (HRTs) comprise a subclass of bHLH proteins that exhibit structural similarity with the HES proteins and include HRT1, HRT2 and HRT3. The HRT family (also designated Hesr, Hey, CHF and Gridlock) contain a bHLH domain, an Orange domain and a novel YRPW domain, which is absent in HRT3. The Hairy-related genes map to human chromosomes 8q21, 6q21 and 1p34.3 for HRT1, HRT2 and HRT3, respectively, and are downstream targets for Notch signaling. HRT1 is expressed in the somitic mesoderm, central nervous system, kidney, heart, nasal epithelium and limb buds in murine embryos as well as in adult tissues. It has altered expression in many breast, lung and kidney tumors. Like HRT1, HRT2 and HRT3 are also expressed in developing somites, heart and nervous system.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HEY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat