- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 36
- Antigen symbol:TRIM36
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:55521
- Antigen synonyms:HAPRIN|RBCC728|RNF98
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
TRIM36 (tripartite motif-containing 36), also known as RNF98 (RING finger protein 98), HAPRIN (haploid germ cell-specific RBCC protein) or RBCC728, is a 728 amino acid protein that belongs to the TRIM/RBCC (Ring finger, B box, coiled-coil) family. Predominantly expressed in prostate, testis and brain with weak expression in heart, kidney and lung, TRIM36 contains two B box-type zinc fingers, a SPRY domain, a coiled-coil domain, a fibronectin type-III domain and a RING-type zinc finger; a motif that has zinc-chelating activity and is involved in mediating protein-protein and protein-DNA interactions. Localizing to the cytoplasm and the acrosomal region of germ cells and mature sperm, TRIM36 is believed to play a role in the acrosome reaction and fertilization. In addition, TRIM36 is overexpressed in prostate cancer, suggesting a possible role for TRIM36 in prostate tumorigenesis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: TRIM36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat