Auftragserfassung
Switzerland
ContactUsLinkComponent
 
Anti-DHPS Rabbit Polyclonal Antibody
Katalognr. BOSSBS-8259R
Anbieter:  Bioss
Anti-DHPS Rabbit Polyclonal Antibody
Katalognr. BOSSBS-8259R
Anbieter:  Bioss

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    deoxyhypusine synthase
  • Antigen-Symbol:
    DHPS
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    1725
  • Antigen-Synonyme:
    Dhps|DS|MIG13|DHYS_HUMAN|Migration inducing gene 13.|DHS|Deoxyhypusine synthase
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Konzentration:
    1 ug/ul
  • Transporttemperatur:
    4 °C
  • Reinigung:
    Purified by Protein A.
  • Größe:
    100 μg
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

Catalyzes the NAD-dependent oxidative cleavage of spermidine and the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DHPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat