Order Entry
Switzerland
ContactUsLinkComponent
Anti-CMTM8 Rabbit Polyclonal Antibody
Anti-CMTM8 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8024R
Supplier:  Bioss
Anti-CMTM8 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8024R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CKLF-like MARVEL transmembrane domain containing 8
  • Antigen symbol:
    CMTM8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    152189
  • Antigen synonyms:
    CKLF8_HUMAN.|CKLF like MARVEL transmembrane domain containing 8|CKLFSF8 V2|Chemokine like factor super family member 8|CMTM 8|Cklfsf8|CMTM8|CMTM-8
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq, Jul 2008].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CMTM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: