Order Entry
Switzerland
ContactUsLinkComponent
Anti-ICMT Rabbit Polyclonal Antibody
Anti-ICMT Rabbit Polyclonal Antibody
Catalog # BOSSBS-8019R
Supplier:  Bioss
Anti-ICMT Rabbit Polyclonal Antibody
Catalog # BOSSBS-8019R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IsoprenylCysteine carboxyl methyltransferase
  • Antigen symbol:
    ICMT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    23463
  • Antigen synonyms:
    PCCMT|Prenylated protein carboxyl methyltransferase|pcCMT|Isoprenylcysteine carboxylmethyltransferase|Prenylcysteine carboxyl methyltransferase|PPMT|Protein-S-isoprenylcysteine O-methyltransferase|ICMT
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the post-translational methylation of isoprenylated C-terminal cysteine residues.Tissue specificity:Ubiquitously expressed. Expressed at higher levels in the cerebellum and putamen than in other brain regions. Abundant expression seen in the Purkinje cells and pontine neurons.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ICMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat