Anti-CK II beta Rabbit Polyclonal Antibody
Katalognr. BOSSBS-7909R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:casein kinase 2, beta polypeptide
- Antigen symbol:CK II beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Gene ID:13001
- Antigen synonyms:CK II beta
- Modification:Ser209
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:200-215/215
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Plays a complex role in regulating the basal catalytic activity of the alpha subunit (By similarity). Participates in Wnt signaling.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CK II beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: