Anti-AdV 5 E1A Rabbit Polyclonal Antibody
BOSSBS-7697R
: Bioss
- Antibody type:Primary
- Antigen name:Adenovirus 5 E1A
- Antigen symbol:AdV 5 E1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:10771
- Antigen synonyms:Protein BS69|Adenovirus 5 E1A-binding protein|MRD30|Bone morphogenetic protein receptor-associated molecule 1|Zinc finger MYND domain-containing protein 11|BRAM1|BS69|ZMYND11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:540-570/602
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Chromatin reader that specifically recognizes and binds histone H3.3 trimethylated at 'Lys-36' (H3.3K36me3) and regulates RNA polymerase II elongation. Does not bind other histone H3 subtypes (H3.1 or H3.2) (By similarity). Colocalizes with highly expressed genes and functions as a transcription corepressor by modulating RNA polymerase II at the elongation stage. Acts as a tumor-suppressor by repressing a transcriptional program essential for tumor cell growth.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: AdV 5 E1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat