Specifications
- Antibody type:Primary
- Antigen name:Nucleolar protein 3
- Antigen symbol:NOL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:8996
- Antigen synonyms:FCM|NOP|Apoptosis repressor with CARD|Myp|MYP|NOP30|Nucleolar protein 3|ARC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD)bcontaining molecules and a caspase family of proteases. CARD containing cell death regulators include RAIDD, RICK, BCL10, Apaf 1, caspase 9 and caspase 2. Apoptosis repressor with CARD is a CARD domain containing protein that interacts with caspase 2 and 8 to inhibit enzymatic activity of caspase 8. Apoptosis repressor with CARD suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR 1, and DR3. The mRNA of Apoptosis repressor with CARD is primarily expressed in skeletal muscle and cardiac tissue. The nuclear isoform (1/Nop30) may be involved in RNA splicing and the cytoplasmic isoform (2/Myp) may inhibit apoptosis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: NOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat