Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-MAGEB2 Rabbit Polyclonal Antibody
Katalognr. BOSSBS-6822R
Anbieter:  Bioss
Anti-MAGEB2 Rabbit Polyclonal Antibody
Katalognr. BOSSBS-6822R
Anbieter:  Bioss

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Melanoma Antigen Family B2
  • Antigen-Symbol:
    MAGEB2
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    4113
  • Antigen-Synonyme:
    CT3.2|MAGE-XP-2|DAM6
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Konzentration:
    1 ug/ul
  • Transporttemperatur:
    4 °C
  • Reinigung:
    Purified by Protein A.
  • Größe:
    100 μg
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21. [provided by RefSeq, Jul 2008].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MAGEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human