Anti-TRIM47 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6744R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 47
- Antigen symbol:TRIM47
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:91107
- Antigen synonyms:RING finger protein 100|Tripartite mot containing protein 47|RNF100|TRI47_HUMAN.|Gene overexpressed in astrocytoma protein|GOA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
TRIM47 belongs to the TRIM/RBCC family. It contains one B box type zinc finger, one B30.2/SPRY domain and one RING type zinc finger. TRIM47 has low expression in most tissues; higher expression in kidney tubular cells and over expressed in astrocytoma tumor cells.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: TRIM47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat