Anti-DDCT Rabbit Polyclonal Antibody
Catalog # BOSSBS-6608R
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:D-dopachrome tautomerase
- Antigen symbol:DDCT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:1652
- Antigen synonyms:D-dopachrome tautomerase|D-dopachrome decarboxylase|DOPD_HUMAN,y Phenylpyruvate tautomerase II.|Ddt|D dopachrome tautomerase|D dopachrome decarboxylase|DDCT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Technische Daten
Über diesen Artikel
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: DDCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat