Anti-HPX Rabbit Polyclonal Antibody
Catalog # BOSSBS-6053R
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:Hemopexin
- Antigen symbol:HPX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:3263
- Antigen synonyms:HX|Beta-1B-glycoprotein|Hemopexin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Technische Daten
Über diesen Artikel
Binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HPX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse