Anti-RPS6KA3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5678R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein S6 Kinase, 90kDa, Polypeptide 3
- Antigen symbol:RPS6KA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:6197
- Antigen synonyms:MRX19|HU-2|ISPK-1|ISPK1|HU-3|RPS6KA3|p90-RSK2|CLS|RSK-2|RSK|RSK2|pp90RSK2|MAPKAPK1B|S6K-alpha3
- Modification:Ser369
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
RSK2 or MAPKAP Kinase 1b is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains two non-identical kinase catalytic domains and phosphorylates various substrates including ribosomal protein S6 and members of the mitogen activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in this gene have been associated with Coffin Lowry syndrome.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: RPS6KA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat