Anti-ICAM1 Rabbit Polyclonal Antibody
BOSSBS-4618R
: Bioss
- Antibody type:Primary
- Antigen name:Intercellular Adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Gene ID:3383
- Antigen synonyms:ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:45-95/532
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). During leukocyte trans-endothelial migration, ICAM1 engagement promotes the assembly of endothelial apical cups through ARHGEF26/SGEF and RHOG activation. In case of rhinovirus infection acts as a cellular receptor for the virus.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human