- Antibody type:Primary
- Antigen name:5' AMP-activated protein kinase alpha
- Antigen symbol:AMPK alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:5562
- Antigen synonyms:ACACA kinase|Hydroxymethylglutaryl-CoA reductase kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|AMPK2|AMPK subunit alpha-2|Acetyl-CoA carboxylase kinase|PRKAA2|AMPK subunit alpha-1|au-protein kinase PRKAA1|PRKAA1|AMPK1|5'-AMP-activated protein kinase catalytic subunit alpha-2|HMGCR kinase
- Modification:Ser496+Ser502
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
The protein encoded by this gene belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5'-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: AMPK alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat