Order Entry
Switzerland
ContactUsLinkComponent
Anti-GPR31 Rabbit Polyclonal Antibody
Anti-GPR31 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13530R
Supplier:  Bioss
Anti-GPR31 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13530R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor 31
  • Antigen symbol:
    GPR31
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    2853
  • Antigen synonyms:
    HETER|HETER1|12-HETER
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interacts with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling, along with involvement in many pathological conditions. GPR31 (G-protein coupled receptor 31) is a 319 amino acid orphan receptor that localizes to the cell membrane. GPR31 shares 25-33% homology with members of the chemokine, purino and somatostatin receptor gene families.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human