Anti-oncomodulin/OCM Rabbit Polyclonal Antibody
BOSSBS-12154R
: Bioss
- Antibody type:Primary
- Antigen name:Oncomodulin
- Antigen symbol:OCM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Antigen synonyms:OCMN|ONCO_HUMAN|Parvalbumin beta.|OM|Oncomodulin-1|Ocm|onco
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Oncomodulin is a high-affinity calcium ion-binding protein. It belongs to the superfamily of calmodulin proteins, also known as the EF-hand proteins. Oncomodulin is an oncodevelopmental protein found in early embryonic cells in the placenta and also in tumors. [provided by RefSeq, Jul 2008].
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: oncomodulin/OCM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat