- Antibody type:Primary
- Antigen name:Acetoacetyl-CoA Synthetase
- Antigen symbol:AACS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:65985
- Antigen synonyms:Acyl-CoA synthetase family member 1|Protein sur-5 homolog|Acetoacetyl-CoA synthetase|AACS_HUMAN.|ACSF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
ACSF1 is a 672 amino acid protein belonging to the ATP-dependent AMP-binding enzyme family. Encoded by a gene that maps to human chromosome 12q24.31, ACSF1 is highly expressed in kidney, heart and brain, and shows similar neural expression as HMGCR (3-hydroxy-3-methylglutaryl-CoA reductase). Existing as three alternatively spliced isoforms, ACSF1 participates in ATP binding, ligase activity, acetoacetate-CoA ligase activity and nucleotide binding. The ACSF1 promoter is a known PPAR?target gene, with the nuclear receptor recruited to the ACSF1 promoter by direct interaction with stimulating protein-1 (Sp1). ACSF1 activates acetoacetate and is highly regulated by modulators that affect HMGCR and cholesterol biosynthesis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: AACS/ACSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat