Order Entry
Switzerland
ContactUsLinkComponent
 
Rabbit IgG Isotype Control (RPE (R-Phycoerythrin))
Catalog # IMMRIR2800
Supplier:  ImmunoReagents
Rabbit IgG Isotype Control (RPE (R-Phycoerythrin))
Catalog # IMMRIR2800
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Goat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage buffer:
    100 nM Sodium Phosphate (pH 7.4), 100 nM NaCl, 50 mM Sucrose, 0.1% BSA and 2 mM sodium azide
  • Storage temperature:
    2-8 °C
  • Shipping temperature:
    2-8 °C
  • Immunogen:
    Goat IgG
  • Tested applications:
    Cell-based assay, Microarray, Microplate applications
  • Purification:
    Affinity purified using solid phase biotin
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

R-phycoerythrin is used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat