Order Entry
Switzerland
ContactUsLinkComponent
NMR sample tubes, SP Wilmad-LabGlass
NMR sample tubes, SP Wilmad-LabGlass
Catalog # WILM516-CC-5
Supplier:  SP Industries
NMR sample tubes, SP Wilmad-LabGlass
Catalog # WILM516-CC-5
Supplier:  SP Industries

Specifications

  • Frequency:
    600 MHz
  • Type:
    Coaxial system, complete set, 5 mm
  • Pk:
    1

Specifications

About this item

Pyrex® glass.

  • 50 µm gap between the inner and outer tubes
  • Ideal for variable temperature studies

These double layered tubes provide protection to toxic samples. They can also be used with a reference standard which is insoluable in the sample or may cause a reaction.