ContactUsLinkComponent
Single gas detectors, BW Clip
Single gas detectors, BW Clip
Supplier:  BW TECHNOLOGIES
Certificates
undefined
Single gas detectors, BW Clip
Supplier:  BW TECHNOLOGIES
Specifications
  • Catalog No:
  • 151-0229
  • 151-0232
  • 151-0228
  • 151-0230
  • 151-0322
  • 118-0484
  • 118-0483
  • 151-0231
  • Humidity range (RH):
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • 5 - 95 %
  • Alarm:
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Visual, vibrating, audible
  • Battery life:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • W×D×H:
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • 41×50×87 mm
  • Weight:
  • 92 g
  • 92 g
  • 92 g
  • 92 g
  • 92 g
  • 92 g
  • 92 g
  • 92 g