Order Entry
Switzerland
ContactUsLinkComponent
 
Hydrometer according to Baumé
Catalog # SCNL3311025
undefined
Hydrometer according to Baumé
Catalog # SCNL3311025

Specifications

  • Description:
    Hydrometer for salt water
  • Length:
    270 mm
  • Division:
    0,5 °Bé
  • Pk:
    1
  • Range:
    0 - 26 °Bé

Specifications

About this item

This hydrometer is specially designed for applications with salt water.

Reference temperature: 20 °C

Ordering information: Available with works or DAkkS calibration certificate.