Orders LinkContactUsLinkComponent
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Column coupler (for use with H0001/H0002)
Catalog # C0001
Supplier:   Avantor
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Catalog # C0001
Supplier:  Avantor
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Column coupler (for use with H0001/H0002)
Price
SFr. 91.40
Log in for availability
1 * 1 ST
Product Details & Documents
Avantor® ACE® PEEK one-piece column couplers are ideal for connecting analytical or semi-preparative columns to stand-alone guard cartridge holders. They can also be used to connect two or more HPLC columns in series or to make similar HPLC connections.
  • Universal connector, ultra-low dead volume
  • Pressure rated to 350 bar
  • Reusable, female-female connector
  • Inert and biocompatible
Specifications
  • Description:
    Column coupler (for use with H0001/H0002)