Order Entry
Switzerland
ContactUsLinkComponent
Di(N-succinimidyl) carbonate for synthesis, Sigma-Aldrich®
Irritant
Di(N-succinimidyl) carbonate for synthesis, Sigma-Aldrich®
Catalog # 8.14971.0005
Supplier:  Merck
CAS Number:  
Irritant
Di(N-succinimidyl) carbonate for synthesis, Sigma-Aldrich®
Catalog # 8.14971.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    188 - 191 °C
  • Molar Mass:
    256.17 g/mol
  • Molecular weight:
    256,17 g/mol
  • Assay (HPLC, area%) :
    ≥98% (a/a)
  • Identity (IR):
    Passes test
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
 :256.17 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:277-730-3
 :GHS07
Storage Temperature:Ambient
CAS Number:74124-79-1
Formula:C₉H₈N₂O₇
MDL Number:MFCD00009767