Orders LinkContactUsLinkComponent
Inspection sets
Inspection sets
Supplier:   Industrial Physics
Inspection sets
Supplier:  Industrial Physics
Certificates
Inspection sets
2 Options Available Below
Product Details & Documents
These basic sets are designed to perform all measurements and tests regarding painting.
Delivery information: Moisture meter, digital thermo/hygrometer, compass, illuminated surface microscope (to check the surface and paint layer), stainless steel cutter knife, pH-indicator paper (to check the acidity of concrete walls and floors), wedge (for checking reverse-side tolerance and thickness of glass), telescopic inspection mirror and lamp (for visual control), waterproof marker and MasterPaintPlate (multifunctional instrument to measure wet film thickness, cover of edges, thickness of glass, as well as providing an indication of the viscosity and adhesion of the coating).
Documents
Product Family Options
Product Information
  • Availability
    Price
  • SIMESP5000
    Inspection set with destructive wood moisture meter
    Inspection set with destructive wood moisture meter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 913.00
    SFr. 913.00
    1 * 1 ST
       SFr. 913.00
  • SIMESP5005
    Inspection set with non-destructive wood moisture meter
    Inspection set with non-destructive wood moisture meter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’080.00
    SFr. 1’080.00
    1 * 1 ST
       SFr. 1’080.00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Knife with rubber grip
    Knife with rubber grip
    SIMESP3008
    Log in for availability
    1 * 1 ST - SFr. 38.80
Product Information
  • Catalog #
    Availability
    Price
  • Sturdy stainless steel bradle for dry rot inspection in timber
    Sturdy stainless steel bradle for dry rot inspection in timber
    SIMESP6202
    Log in for availability
    1 * 1 ST - SFr. 94.50
Product Information
  • Catalog #
    Availability
    Price
  • Water resistant marker
    Water resistant marker
    SIMESP6400
    Log in for availability
    1 * 1 ST - SFr. 17.10