Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Llama
- Reactivity:Rabbit,Sheep,Human,Mouse
- Immunogen:purified rabbit IgG, whole molecule
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Llama anti-Rabbit IgG (H&L), DyLight 488 Conjugate is a secondary antibody which binds to heavy (?) chains chains on rabbit IgG light chains on all rabbit immunoglobulins.DyLight® 488 Amax = 493 nm, Emax = 518 nm. Antibodies are purified using solid phase Rabbit IgG (H&L).DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Llama anti-Rabbit IgG (H&L), DyLight 488 Conjugate is a secondary antibody which binds to heavy (?) chains chains on rabbit IgG light chains on all rabbit immunoglobulins.DyLight® 488 Amax = 493 nm, Emax = 518 nm. Antibodies are purified using solid phase Rabbit IgG (H&L).DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Llama
Isotype:
Reactivity: Human, Mouse, Rabbit, Sheep