Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-PsbA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-PsbA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # AGRIAS05084A-HRP
Supplier:  Agrisera
undefined
Anti-PsbA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # AGRIAS05084A-HRP
Supplier:  Agrisera

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    PSBA
  • Klonalität:
    Polyklonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • Wirt:
    Rabbit
  • Reaktivität:
    Zea mays,
    A. thaliana,
    H. vulgare
  • Western blot:
    Yes
  • UniProtKB:
    A4QJR4
  • Immunogen:
    KLH-conjugated synthetic peptide derived from available plant, algal and cyanobacterial PsbA sequences, including Arabidopsis thaliana
  • Größe:
    40 µg
  • VE:
    40 µG

Specifications

About this item

The psbA gene has been cloned from many species of plants, green algae, and cyanobacteria. The psbA gene is located in the chloroplast genome and encodes for the D1 protein, a core component of Photosystem II. PsbA/D1 is rapidly cycled under illumination in all oxygenic photobionts. Tracking PsbA pools using the Global PsbA antibody can show the functional content of Photosystem II in a wide range of samples. Alternative names: 32 kDa thylakoid membrane protein, photosystem II protein D1.,The psbA gene has been cloned from many species of plants, green algae, and cyanobacteria. The psbA gene is located in the chloroplast genome and encodes for the D1 protein, a core component of Photosystem II. PsbA/D1 is rapidly cycled under illumination in all oxygenic photobionts. Tracking PsbA pools using the Global PsbA antibody can show the functional content of Photosystem II in a wide range of samples. Alternative names: 32 kDa thylakoid membrane protein, photosystem II protein D1.

Type: Primary
Antigen: PSBA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Zea mays