F(ab)'2 Anti-alpha Fetoprotein (AFP) Goat Polyclonal Antibody
Catalog # IMMRIR2914
Supplier: ImmunoReagents
Specifications
- Antibody type:Primary
- Antigen name:Alpha-fetoprotein
- Antigen symbol:AFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid, 0.2 µm filtered
- Antigen synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Antibody fragment:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:2-8 °C
- Concentration:>5 mg/ml
- Shipping temperature:2-8 °C
- Immunogen:Human Alpha Fetoprotein (AFP)
- Purification:Proprietary 2-step fractionation procedure
- Size:1,0 mg
- Pk:1 mg
Specifications
About this item
This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications
Type: Primary
Antigen: Alpha Fetoprotein (AFP)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human