- Antigen name:Histidine triad nucleotide binding protein 1
- Antigen symbol:HINT1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:3094
- Antigen synonyms:NMAN|PRKCNH1|PKCI-1|HINT
- UniProtKB:P49773
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human HINT1 (59-92aa HISQISVAEDDDESLLGHLMIVGKKCAADLGLNK), different from the related mouse and rat sequences by three amino acids.
- Purification:Immunogen affinity purified.
- Size:100 μg / vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for Histidine triad nucleotide-binding protein 1(HINT1) detection. Tested with WB,IHC-P in Human;Mouse;Rat.
Histidine triad nucleotide-binding protein 1 is a protein that in humans is encoded by the HINT1 gene. It is a haploinsufficient tumor suppressor gene that inhibits the Wnt/?-catenin pathway in colon cancer cells and Microphthalmia-associated transcription factor (MITF) activity in human mast cells. The protein encoded by this gene can hydrolyze substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. Several transcript variants, but only one of them protein-coding, have been found for this gene.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.