ContactUsLinkComponent
Reagents for non Merck photometers, Spectroquant®, Supelco®
Reagents for non Merck photometers, Spectroquant®, Supelco®
Catalog # 1.19254.0001
Supplier:  Merck
CAS Number:  
undefined
Reagents for non Merck photometers, Spectroquant®, Supelco®
Catalog # 1.19254.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Chlorine powder packs (10 ml free chlorine samples)
  • Seq:
    01
  • Tests per kit:
    100
  • Range:
    0 - 2,00 mg/l
Specifications
About this item
Spectroquant® test kits are suitable for use only in conjunction with Hach instruments. They do not require any further device-specific calibration.