Order Entry
Switzerland
ContactUsLinkComponent
Magnesia, rods Ø×H 2×140 mm, for the phosphorus salt pearls, Supelco®
Magnesia, rods Ø×H 2×140 mm, for the phosphorus salt pearls, Supelco®
Catalog # 1.05809.0100
Supplier:  Merck
CAS Number:  
undefined
Magnesia, rods Ø×H 2×140 mm, for the phosphorus salt pearls, Supelco®
Catalog # 1.05809.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0100

Specifications

Specification Test Results

  • Length:
    about 14 cm
  • Diameter:
    about 2 mm
  • Flame test:
    Passes test
  • Pearl test:
    Passes test

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
 :1309-48-4