ContactUsLinkComponent
Transfer cart for glassware washing machines
Transfer cart for glassware washing machines
Catalog # 141-0510
Supplier:  MIELE
undefined
Transfer cart for glassware washing machines
Catalog # 141-0510
Supplier:  MIELE
Specifications
  • Description:
    MF/3 Transfer cart for glassware washing machine, Miele G 7823, G 7824, G 7825, G 7826
  • W×D×H:
    660×807× 1182 (+/- 100 mm) mm
  • Pk:
    1
  • Shelves:
    2
Specifications
About this item
Transfer cart for loading and unloading of wash carts.
  • Glassware is loaded/unloaded outside of machine allowing sufficient space for an easy loading/unloading compared to loading directly inside the wash chamber
  • Drip tray to capture liquid dripping from contaminated glassware
  • Foot-switch operated height-adjustment mechanism
  • With four lockable wheels