Orders LinkContactUsLinkComponent
Bovine serum
Bovine serum
Normal bovine serum, Lyophilised
Catalog # IMMRIR2420
Supplier:   ImmunoReagents
Bovine serum
Catalog # IMMRIR2420
Supplier:  ImmunoReagents
Bovine serum
Bovine serum
Normal bovine serum, Lyophilised
Price
SFr. 193.00
Log in for availability
1 * 10 mL
Product Details & Documents
Normal bovine serum is used as a blocking agent or as a control for immunoassays. All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer.
NOTE: All ImmunoReagents products are provided with a standard product CoA which includes application, purity and cross reactivity profiles. Additionally, products can by customised based on specific assay requirements, please contact your local Avantor sales office for more information.
Certifications: (EC)142/2011 approved by APHIS, Reference Number NC-TEC-0002
Delivery information: Serum is supplied in lyophilised form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice or to aliquot reconstituted serum and store frozen to prevent contamination.
Specifications
  • Serum format:
    Lyophilised
  • Description:
    Normal bovine serum