Stopcocks for VWR® Collection Carboys
216-1745
- Description:Stopcock, with adapter for tubing connection and stopper for long storage for VWR carboys
- Pk:1
PP, white/purple.
Pack consists of stopcock, adapter for tube connection and stopper for long storage. Stopcock is designed to be tightly assembled by hand and, therefore, does not require any tools for assembly. Autoclavable.
Resin meets US FDA 21 CFR, USP Class VI