ContactUsLinkComponent
Straw pipettes, STRAWS 180
Straw pipettes, STRAWS 180
Catalog # GRADSTW180
Supplier:  Grade Products
Straw pipettes, STRAWS 180
Catalog # GRADSTW180
Supplier:  Grade Products
Specifications
  • Packed:
    50 straws/sachet, 40 sachets/case
  • Description:
    Straw pipettes
  • Pk:
    2.000
Specifications
About this item
These disposable pipette straws are specifically designed for use with laboratory blender bags. They are sterile, unbreakable and eliminate the hazard of broken glass. They also avoid exposure to infectious materials and contaminating samples.
Length: 180 mm
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904