Impact testers
Supplier: Industrial Physics
Jump to:
About this item
Impact testers used to determine the impact resistivity and flexibility of materials.
- Spirit level built-in for correct positioning
- Dual scale instrument equipped with a special guidance
- Distance between each impact is always according to the standards
: DIN 6272-1 and -2, ASTM D2794 and G14.
2 Options Available Below
Product Details & Documents
Impact testers used to determine the impact resistivity and flexibility of materials.
- Spirit level built-in for correct positioning
- Dual scale instrument equipped with a special guidance
- Distance between each impact is always according to the standards
: DIN 6272-1 and -2, ASTM D2794 and G14.
Documents
TQC Company Video
Videos
Product Family Options
Product Information
- AvailabilityPrice
- SIMESP1890Impact tester, ISO 6272-1, incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm and die 27 mm, 1 kgImpact tester, ISO 6272-1, incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm and die 27 mm, 1 kgLog in for availability1 * 1 STSFr. 3’020.00
- SIMESP1891Dual impact tester, ISO 6272-1 and ASTM D2794 (before 1993), incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm, die 27 mm, 1 kg, ball Ø 15,9 mm and die 16,3 mm, 0,9 kgDual impact tester, ISO 6272-1 and ASTM D2794 (before 1993), incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm, die 27 mm, 1 kg, ball Ø 15,9 mm and die 16,3 mm, 0,9 kgLog in for availability1 * 1 STSFr. 3’330.00