Order Entry
Switzerland
ContactUsLinkComponent
Qualitative filter papers, grades MN 619 de and 619 de 1/4, circles and folded filters
Qualitative filter papers, grades MN 619 de and 619 de 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Qualitative filter papers, grades MN 619 de and 619 de 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL

Specifications

  • Catalog No:
  • 516-1170
  • MANA439018
  • MANA439009
  • MANA539015
  • MANA539018
  • MANA539011
  • MANA439032
  • MANA539012
  • MANA539032
  • MANA439024
  • MANA539024
  • MANA439007
  • MANA539009
  • MANA439005
  • MANA439015
  • MANA539005
  • MANA439011
  • MANA539007
  • Weight:
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • Thickness:
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • Particle retention:
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • Filtration speed:
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • Ash content:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%

Specifications

Frequently Bought Together