Orders LinkContactUsLinkComponent
Permanent marker
Permanent marker
Permanent marker
Certificates
Permanent marker
3 Options Available Below
Product Details & Documents
This versatile paint marker dispense a fast-drying, high-gloss enamel paint for permanent marking, lettering, coding and numbering of both interior and exterior surfaces of metal, plastic, printed circuit boards, glass, wood, paper, leather, textiles, etc. This marker can write on porous or non-porous; rough or smooth; wet, oily or dry surfaces.
  • Marks are permanent
  • Withstand heat and weathering under adverse conditions
  • Available in different lead-free colours
  • With long-lasting replaceable fiber tips
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • KOEH2-5064
    Permanent marker, fiber tip
    Permanent marker, fiber tip
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31.80 SFr.
    31.80 SFr.
    1 * 1 ST
       31.80 SFr.
  • KOEH2-5068
    Permanent marker, fiber tip
    Permanent marker, fiber tip
    Silver
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31.30 SFr.
    31.30 SFr.
    1 * 1 ST
       31.30 SFr.
  • KOEH2-5071
    Permanent marker, fiber tip
    Permanent marker, fiber tip
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32.70 SFr.
    32.70 SFr.
    1 * 1 ST
       32.70 SFr.