ContactUsLinkComponent
Gas detection tubes for nitrous fumes
Gas detection tubes for nitrous fumes
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for nitrous fumes
Supplier:  Dräger
Certificates
Specifications
  • Catalog No:
  • 111-0235
  • DRAE8103661
  • Model:
  • Nitrous fumes 2/a
  • Nitrous fumes 0.2/a
  • Colour change:
  • Yellow to blue grey
  • Greyish green to blue grey
  • Humidity:
  • ≤ 30 mg/l H₂O
  • ≤ 40 mg/l H₂O
  • Measurement time:
  • 1/2 minutes
  • 75/30 seconds
  • No. of strokes:
  • 5/10
  • 5/2
  • Standard deviation:
  • ±10 - 15 %
  • ±10 - 15 %
  • Standard range:
  • 5 - 100/2 - 50 ppm
  • 0,2 - 6 ppm
  • Temperature range:
  • 10…30 °C
  • 10…40 °C