Gas detection tubes for alcohols
Catalog # DRAE8103761
Supplier: Dräger
Specifications
- Environmentally Preferable:
- Standard range:-
- Standard deviation:±30 %
- No. of strokes:6
- Colour change:Yellow to mint green
- Temperature range:-
- Humidity:-
- Measurement time:1,5 minutes
- Parameter(s):Ethanol 100/a
- Pk:10
- Range:100 - 3000 ppm
Specifications
About this item
To measure the 'spot concentrations' of alcohol for use with an Accuro pump.
Cross sensitivity Alcohol 100/a: The tube does not differentiate between different alcohols. Higher molecular weight alcohols are indicated with a markedly reduced sensitivity. Aldehydes, ethers, ketones, esters are indicated but with different sensitivity. Aromates, petroleum, halogenated hydrocarbons are not indicated.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.