Anti-FUCA1 Antibody Pair
ABNOH00002517-AP51
: Abnova
- Antigen name:Fucosidase, alpha-L- 1, tissue
- Antigen symbol:FUCA1
- Host:Multiple
- Reactivity:Human
- Gene ID:2517
- Antigen synonyms:FUCA
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human FUCA1.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-FUCA1 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-FUCA1 (50 µg);
Type:
Antigen: FUCA1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human