Orders LinkContactUsLinkComponent
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
   Sustainable Options Available
Supplier:   Avantor
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
Supplier:  Avantor
   Sustainable Options Available
Certificates
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
9 Options Available Below
Product Details & Documents
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Maximum recommended pH: 2,0 to 8,0 (1,5 to 11,5 for SuperC18)
Max. pressure: 1000 bar
Delivery information: Avantor® ACE® Extended Method Development Kits contain Avantor® ACE® SuperC18™, Avantor® ACE® C18-Amide and Avantor® ACE® CN-ES stainless steel columns.
Specifications for Product Family
Specifications
  • Catalog No:
  • MDKE-2-0502U
  • MDKE-2-1002U
  • MDKE-2-1502U
  • MDKE-2-0503U
  • MDKE-2-1003U
  • MDKE-2-1503U
  • MDKE-2-0546U
  • MDKE-2-1046U
  • MDKE-2-1546U
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • MDKE-2-0502U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’010.00
    SFr. 1’010.00
    1 * 1 KIT
       SFr. 1’010.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-0502U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1002U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’260.00
    SFr. 1’260.00
    1 * 1 KIT
       SFr. 1’260.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1002U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1502U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’400.00
    SFr. 1’400.00
    1 * 1 KIT
       SFr. 1’400.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1502U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-0503U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    3,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-0503U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1003U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    3,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’330.00
    SFr. 1’330.00
    1 * 1 KIT
       SFr. 1’330.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1003U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1503U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    3,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’580.00
    SFr. 1’580.00
    1 * 1 KIT
       SFr. 1’580.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1503U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-0546U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’260.00
    SFr. 1’260.00
    1 * 1 KIT
       SFr. 1’260.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-0546U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1046U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’750.00
    SFr. 1’750.00
    1 * 1 KIT
       SFr. 1’750.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1046U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • MDKE-2-1546U
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Extended MDK, SuperC18™, C18-Amide, CN-ES
    Method development kit
    90/100 Å
    2 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 2’820.00
    SFr. 2’820.00
    1 * 1 KIT
       SFr. 2’820.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKE-2-1546U
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact