Order Entry
Switzerland
ContactUsLinkComponent
Analogue stopwatch
Analogue stopwatch
Catalog # HANH121.0117-00
Supplier:  HANHART
undefined
Analogue stopwatch
Catalog # HANH121.0117-00
Supplier:  HANHART

Specifications

  • Ø:
    55 mm
  • Colour:
    Black
  • Description:
    Analogue stopwatch, 1/5 s, 30 min, 80 g
  • Pk:
    1

Specifications

About this item

Addition timer in functional ABS case.

  • Pin lever, 1 jewel
  • Timing capacity: 30 minutes
  • Very lightweight and easy to handle
  • Shock protected