Orders LinkContactUsLinkComponent
Digital hotplates, Cimarec+™
Digital hotplates, Cimarec+™
Digital hotplates, Cimarec+™
Certificates
Digital hotplates, Cimarec+™
Digital hotplates, Cimarec+™
3 Options Available Below
Product Details & Documents
These hotplates feature a raised display design which protects the electronics from any liquid spills.
  • Easy-to-read LED display for heating temperature
  • Hot top warning indicator and automatic over-temperature protection
  • Temperature adjustable in 1 degree increments
  • Top surface temperature could rise to the maximum
  • 12 litre capacity
  • Heating up time is 8 min to maximum temperature
Temperature control accuracy (at 37 °C): ≤±5 °C
Temperature uniformity (at 37 °C): ≤±2 °C
Delivery information: Supplied with cCSAUS certificate. Other accessories such as optional PT1000 temperature probe and splash guard protection shield are available, these must be ordered separately
Product Family Options
Product Information
  • Heating area
    Temp.
    Plate size
    Type
    W×D×H
    Availability
    Price
  • 108×108 mm
    30...540 °C
    108×108 mm
    Hotplate, Cimarec+
    127×254×91 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 360.00
    SFr. 360.00
    1 * 1 ST
       SFr. 360.00
  • 184×184 mm
    30...540 °C
    184×184 mm
    Hotplate, Cimarec+
    208×330×97 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 439.00
    SFr. 439.00
    1 * 1 ST
       SFr. 439.00
  • 260×260 mm
    30...400 °C
    260×260 mm
    Hotplate, Cimarec+
    287×411×102 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 664.00
    SFr. 664.00
    1 * 1 ST
       SFr. 664.00
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • PT1000 external probe, stainless steel, 0 - 400 °C
    PT1000 external probe, stainless steel, 0 - 400 °C
    BARNCIC0001448
    Cimarec™ and SuperNuova™ hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 83.90
Product Information
  • Catalog #
    For
    Availability
    Price
  • PT100 external probe, stainless steel, 0 - 400 °C
    PT100 external probe, stainless steel, 0 - 400 °C
    BARNCIC0001447
    Cimarec™ and SuperNuova™ hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 78.50
Product Information
  • Catalog #
    For
    Availability
    Price
  • Splash guard/protection shield, transparent
    Splash guard/protection shield, transparent
    BARNCIC0001446
    10×10'' Cimarec™ and SuperNuova™ stirrers, hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 32.20
Product Information
  • Catalog #
    For
    Availability
    Price
  • Splash guard/protection shield, transparent
    Splash guard/protection shield, transparent
    BARNCIC0001445
    7×7'' Cimarec™ and SuperNuova™ stirrers, hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 26.10
Product Information
  • Catalog #
    For
    Availability
    Price
  • PT1000 external probe, Hastelloy®, 0 - 400 °C
    PT1000 external probe, Hastelloy®, 0 - 400 °C
    BARNCIC0001449
    Cimarec™ and SuperNuova™ hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 125.00
Product Information
  • Catalog #
    For
    Availability
    Price
  • Splash guard/protection shield, transparent
    Splash guard/protection shield, transparent
    BARNCIC0001444
    4×4'' Cimarec™ and SuperNuova™ stirrers, hotplates and hotplate stirrers
    Log in for availability
    1 * 1 ST - SFr. 30.30