Orders LinkContactUsLinkComponent
Circular spacers
Supplier:   PerkinElmer
Circular spacers
Supplier:  PerkinElmer
Certificates
Circular spacers
4 Options Available Below
Product Details & Documents
These circular spacers are designed to be used for the analysis of less viscous samples.
  • Available in 0,025 to 0,500 mm thickness
Since the demountable cells uses the circular windows, circular spacers should be used.
Product Family Options
Product Information
  • For
    Availability
    Price
  • PERC51001135
    Circular Teflon spacers, 0,05 mm
    Circular Teflon spacers, 0,05 mm
    FTIR, 1600 series
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 210.00
    SFr. 210.00
    1 * 5 ST
       SFr. 210.00
  • PERC51001137
    Circular Teflon spacers, 0,01 mm
    Circular Teflon spacers, 0,01 mm
    FTIR, 1600 series
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 210.00
    SFr. 210.00
    1 * 5 ST
       SFr. 210.00
  • PERC51001140
    Circular Teflon spacers, 0,2 mm
    Circular Teflon spacers, 0,2 mm
    FTIR, 1600 series
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 210.00
    SFr. 210.00
    1 * 3 ST
       SFr. 210.00
  • PERC51001141
    Circular Teflon spacers, 0,5 mm
    Circular Teflon spacers, 0,5 mm
    FTIR, 1600 series
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 242.00
    SFr. 242.00
    1 * 3 ST
       SFr. 242.00