Orders LinkContactUsLinkComponent
DNA gel loading buffer 6X
DNA gel loading buffer 6X
6X DNA loading buffer, 5× 1 ml, 5
Catalog # LONZ50655
Supplier:   LONZA
DNA gel loading buffer 6X
Catalog # LONZ50655
Supplier:  LONZA
CAS Number:  
DNA gel loading buffer 6X
DNA gel loading buffer 6X
6X DNA loading buffer, 5× 1 ml, 5
CAS Number:  
Price
122.00 SFr.
Log in for availability
1 * 5 ST
Product Details & Documents
This buffer is a Ficoll based loading buffer containing Ficoll®, EDTA, xylene cyanol, and bromophenol blue. Ideal for separation of DNA fragments with improved resolution.
Specifications
Specifications
  • Pk:
    5
  • Description:
    6X DNA loading buffer, 5× 1 ml
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • 6X DNA loading buffer, 5× 1 ml
    6X DNA loading buffer, 5× 1 ml
    LONZ50655
    5
    Log in for availability
    1 * 5 ST - SFr. 122.00