ContactUsLinkComponent
Calibration gas cylinders
Calibration gas cylinders
Catalog # 111-1703
Supplier:  M S A
CAS Number:  
undefined
Calibration gas cylinders
Catalog # 111-1703
Supplier:  M S A
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Calibration gas cylinder (58 litres of gas): 1,45% CH4; 20% H2S; 60% CO; 15% O2
Specifications
About this item
The gas mixture in calibration gas cylinder is certified to be prepared gravimetrically, using NIST traceable weights. The lot number and nominal value of the gas constituents in volume, percent by mass, PPM, or volume is specified on the cylinder.