Temperature measuring for urine
Catalog # 700-9660
Supplier: SURESCREEN
Specifications
- Description:Temperature strip
- Environmentally Preferable:
- Pk:100
Specifications
About this item
For use with the standard collection cup, and used to show if a urine sample is within range of body temperature.
- Samples outside this temperature range could have been adulterated